Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK089086.1 | - | 15 | 1592 | ||
AK089169.1 | - | 15 | 1614 | |||
AK134118.1 | - | 5 | 1146 | |||
AK150411.1 | - | 15 | 1594 | |||
AK151535.1 | - | 7 | 823 | |||
AK152675.1 | - | 15 | 1611 | |||
AK152769.1 | - | 15 | 1593 | |||
AK153065.1 | - | 15 | 1593 | |||
AK155772.1 | - | 15 | 1592 | |||
AK155912.1 | - | 15 | 1612 | |||
AK160041.1 | - | 15 | 1595 | |||
AK170019.1 | - | 15 | 1617 | |||
AK171022.1 | - | 14 | 1905 | |||
AK171060.1 | - | 15 | 1592 | |||
BC025584.1 | - | 15 | 1646 | 5324943 | ||
X13335.1 | - | 15 | 1591 |