Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Add2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271857.1 | NP_001258786.1 | 13 | 1922 | ||
NM_001271858.1 | NP_001258787.1 | 13 | 1919 | |||
NM_001271859.1 | NP_001258788.1 | 13 | 1720 | |||
NM_001271860.1 | NP_001258789.1 | 13 | 1922 | |||
NM_001271861.1 | NP_001258790.1 | 13 | 1723 | |||
NM_013458.5 | NP_038486.2 | 13 | 1723 | |||
GenBank mRNA | AF100422.1 | - | 12 | 1598 | ||
AF100423.1 | - | 12 | 1598 | |||
AF189769.1 | - | 11 | 1520 | |||
AK014496.1 | - | 13 | 1688 | |||
AK028425.1 | - | 13 | 1694 | |||
AK029488.1 | - | 13 | 1873 | |||
BC046783.1 | - | 13 | 1856 | 6401200 | ||
BC053032.1 | - | 13 | 1856 | 6401200 |