Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000841.3 | NP_000832.1 | 8, 9 | 2000 | ||
NM_001256809.2 | NP_001243738.1 | 6, 7 | 1383 | |||
NM_001256811.2 | NP_001243740.1 | 7, 8 | 1859 | |||
NM_001256812.2 | NP_001243741.1 | 7, 8 | 1084 | |||
NM_001256813.2 | NP_001243742.1 | 8, 9 | 1308 | |||
NM_001282847.1 | NP_001269776.1 | 7, 8 | 1783 | |||
XM_017010790.1 | XP_016866279.1 | 8, 9 | 1938 | |||
XM_017010791.1 | XP_016866280.1 | 8, 9 | 1993 | |||
XM_017010792.1 | XP_016866281.1 | 7, 8 | 1537 | |||
XM_017010793.1 | XP_016866282.1 | 7, 8 | 1288 | |||
GenBank mRNA | AK122836.1 | - | 5, 6 | 1974 | ||
AK122982.1 | - | 8, 9 | 1293 | |||
AK126746.1 | - | 6, 7 | 1053 | |||
AK131536.1 | - | 6, 7 | 1383 | |||
AK289524.1 | - | 2318 | ||||
AK289534.1 | - | 8, 9 | 1943 | |||
AK289563.1 | - | 6, 7 | 857 | |||
AK293913.1 | - | 7, 8 | 1084 | |||
AK293949.1 | - | 7, 8 | 1110 | |||
AK312300.1 | - | 7, 8 | 1738 | |||
BC130526.1 | - | 7, 8 | 1658 | 40146557 | ||
BC130528.1 | - | 7, 8 | 1658 | 40146559 | ||
BC144057.1 | - | 6, 7 | 1517 | 9052577 | ||
EU432124.1 | - | 7, 8 | 1499 | |||
U92457.1 | - | 7, 8 | 1524 | |||
X80818.1 | - | 7, 8 | 1669 |
Set Membership: |
G-protein coupled receptor |