Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009626.4 | NP_033756.2 | 3 | 314 | ||
GenBank mRNA | AF331802.1 | - | 3 | 312 | ||
AF331803.1 | - | 3 | 312 | |||
AK009588.2 | - | 3 | 281 | |||
AK009600.1 | - | 3 | 327 | |||
AK144795.1 | - | 265 | ||||
BC047267.1 | - | 3 | 243 | 6310424 | ||
BQ560660.1 | - | 3 | 314 | |||
U20257.1 | - | 3 | 205 |