Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243041.1 | NP_001229970.1 | 8, 9 | 863 | ||
NM_134079.4 | NP_598840.1 | 8, 9 | 820 | |||
GenBank mRNA | AK050135.1 | - | 2, 3 | 123 | ||
AK151863.1 | - | 8, 9 | 865 | |||
AK160264.1 | - | 8, 9 | 817 | |||
AY540996.1 | - | 8, 9 | 762 | |||
AY540997.1 | - | 7, 8 | 707 | |||
BC009659.1 | - | 8, 9 | 785 | 3485959 |