Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr182 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007412.2 | NP_031438.2 | 2 | 827 | ||
GenBank mRNA | AK031480.1 | - | 762 | |||
BC125279.1 | - | 2 | 698 | |||
D17292.1 | - | 2 | 722 | |||
L20338.1 | - | 1 | 168 |
Set Membership: |
G-protein coupled receptor |