Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adora2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009630.3 | NP_033760.2 | 2 | 639 | ||
GenBank mRNA | AK206618.1 | - | 1 | 28 | ||
BC110692.1 | - | 1 | 139 | 30242398 | ||
CJ135665.1 | - | 2 | 573 | |||
CX241487.1 | - | 2 | 444 | |||
U05672.1 | - | 1 | 364 |
Set Membership: |
G-protein coupled receptor |