Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000843.3 | NP_000834.2 | 9 | 2356 | ||
GenBank mRNA | BX647168.1 | - | 1 | 900 |
Set Membership: |
G-protein coupled receptor |