Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005847.2 | NP_001005847.1 | 9 | 1084 | ||
NM_001205054.1 | NP_001191983.1 | 9 | 1054 | |||
GenBank mRNA | BC048230.1 | - | 10 | 2045 | ||
BC132227.1 | - | 9 | 1059 | 40130504 | ||
BC132229.1 | - | 9 | 1059 | 40130506 | ||
BC145297.1 | - | 9 | 1029 | 9053851 | ||
BE656262.1 | - | 5 | 483 | |||
S81393.1 | - | 9 | 1001 |