Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005847.2 | NP_001005847.1 | 6, 7 | 798 | ||
GenBank mRNA | BC048230.1 | - | 7, 8 | 1759 | ||
BC132227.1 | - | 6, 7 | 773 | 40130504 | ||
BC132229.1 | - | 6, 7 | 773 | 40130506 | ||
BE656262.1 | - | 2, 3 | 197 | |||
BQ571087.1 | - | 6, 7 | 690 | |||
S81393.1 | - | 6, 7 | 715 |