Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ager | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271422.1 | NP_001258351.1 | 6 | 589 | ||
NM_001271423.1 | NP_001258352.1 | 6 | 562 | |||
NM_001271424.1 | NP_001258353.1 | 5 | 684 | |||
NM_007425.3 | NP_031451.2 | 6 | 589 | |||
GenBank mRNA | AB207883.1 | - | 6 | 549 | ||
AK004802.1 | - | 6 | 577 | |||
BC061182.1 | - | 6 | 577 | 30300427 | ||
EU520325.1 | - | 6 | 545 | |||
EU570240.1 | - | 6 | 545 | |||
EU570241.1 | - | 5 | 740 | |||
EU570242.1 | - | 6 | 545 | |||
EU570243.1 | - | 6 | 545 | |||
EU570244.1 | - | 5 | 740 | |||
EU570245.1 | - | 5 | 740 | |||
EU570246.1 | - | 5 | 702 | |||
EU570247.1 | - | 5 | 740 | |||
EU906862.1 | - | 6 | 545 | |||
EU906863.1 | - | 6 | 518 | |||
EU906864.1 | - | 5 | 740 | |||
GU164727.1 | - | 5 | 659 | |||
GU164728.1 | - | 6 | 564 | |||
GU164729.1 | - | 6 | 564 | |||
GU164730.1 | - | 6 | 564 | |||
GU164731.1 | - | 6 | 564 | |||
GU164732.1 | - | 6 | 564 | |||
KC692918.1 | - | 6 | 545 | |||
L33412.1 | - | 6 | 550 |