Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Angpt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007426.4 | NP_031452.2 | 9 | 1689 | ||
GenBank mRNA | AF004326.1 | - | 9 | 1615 | ||
AK019860.1 | - | 9 | 1678 | |||
AK048622.1 | - | 9 | 1646 | |||
AK143974.1 | - | 9 | 1677 | |||
AK156132.1 | - | 9 | 1660 | |||
BC027216.1 | - | 9 | 1642 | 3494566 | ||
BC138309.1 | - | 9 | 1583 | 8861329 | ||
BC138313.1 | - | 9 | 1583 | 8861333 |