Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000845.2 | NP_000836.2 | 1 | 564 | ||
NM_001127323.1 | NP_001120795.1 | 2 | 810 | |||
NR_028041.1 | 1 | 564 | ||||
XM_006715938.3 | XP_006716001.1 | 2 | 884 | |||
XM_011516091.1 | XP_011514393.1 | 2 | 1062 | |||
XM_011516092.2 | XP_011514394.1 | 2 | 1062 | |||
XM_017012074.1 | XP_016867563.1 | 2 | 663 | |||
GenBank mRNA | AJ236921.1 | - | 1 | 268 | ||
AJ236922.1 | - | 1 | 268 | |||
AK290197.1 | - | 2 | 861 | |||
AK315203.1 | - | 2 | 810 | |||
AY608335.1 | - | 1 | 253 | |||
BC093725.1 | - | 1 | 279 | 7939570 | ||
BC101675.1 | - | 1 | 279 | 8069181 | ||
BC143344.1 | - | 1 | 279 | |||
EU432125.1 | - | 1 | 253 | |||
U92459.1 | - | 1 | 310 | |||
U95025.1 | - | 1 | 253 |
Set Membership: |
G-protein coupled receptor |