Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002089.3 | NP_002080.1 | 4 | 730 | ||
GenBank mRNA | BC015753.1 | - | 4 | 638 | 4875789 | |
BX334969.2 | - | 4 | 740 | |||
M36820.1 | - | 4 | 642 | |||
M57731.1 | - | 4 | 641 | |||
X53799.1 | - | 4 | 612 |