Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033966.4 | NP_001029138.1 | 5, 6 | 614 | ||
NM_016895.4 | NP_058591.2 | 5, 6 | 614 | |||
GenBank mRNA | AB020202.1 | - | 5, 6 | 536 | ||
AK010936.1 | - | 5, 6 | 533 | |||
AK010951.1 | - | 5, 6 | 535 | |||
AK050133.1 | - | 5, 6 | 575 | |||
AK077519.1 | - | 5, 6 | 537 | |||
AK166976.1 | - | 5, 6 | 525 | |||
AK168056.1 | - | 5, 6 | 537 | |||
AK168148.1 | - | 5, 6 | 564 | |||
BC008610.1 | - | 5, 6 | 524 | 3584786 |