Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ak4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177602.1 | NP_001171073.1 | 4 | 643 | ||
NM_001177604.1 | NP_001171075.1 | 4 | 456 | |||
NM_001177605.1 | NP_001171076.1 | 4 | 434 | |||
NM_009647.5 | NP_033777.1 | 3 | 401 | |||
GenBank mRNA | AB020239.1 | - | 3 | 365 | ||
AK134600.1 | - | 4 | 646 | |||
AK144439.1 | - | 3 | 371 | |||
AK154374.1 | - | 4 | 460 | |||
AK166975.1 | - | 3 | 323 | |||
BC086663.1 | - | 3 | 349 | 30649610 | ||
BC086791.1 | - | 3 | 365 | |||
BY740484.1 | - | 4 | 435 | |||
D85036.1 | - | 3 | 365 |