Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287172.1 | NP_001274101.1 | 8 | 1075 | ||
NM_001287176.1 | NP_001274105.1 | 8 | 1198 | |||
NM_007431.3 | NP_031457.2 | 8 | 1050 | |||
GenBank mRNA | AB473959.1 | - | 7 | 814 | ||
AK076459.1 | - | 8 | 985 | |||
AK161780.1 | - | 8 | 986 | |||
AK164002.1 | - | 8 | 1043 | |||
AK166732.1 | - | 8 | 984 | |||
AK167023.1 | - | 8 | 985 | |||
AK167481.1 | - | 8 | 985 | |||
BC065175.1 | - | 8 | 973 | 6807509 | ||
J02980.1 | - | 8 | 989 | |||
X13409.1 | - | 8 | 933 |