Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alas2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102446.1 | NP_001095916.1 | 11 | 1728 | ||
NM_009653.3 | NP_033783.1 | 11 | 1773 | |||
GenBank mRNA | AK002642.1 | - | 11 | 1744 | ||
AK077610.1 | - | 11 | 1745 | |||
AK165257.1 | - | 11 | 1736 | |||
AK165262.1 | - | 11 | 1748 | |||
AK172094.1 | - | 11 | 1737 | |||
BC024575.1 | - | 7 | 1141 | |||
BC150870.1 | - | 11 | 1720 | 9087781 | ||
M15268.1 | - | 11 | 1686 | |||
M63244.1 | - | 11 | 1743 |