Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009654.3 | NP_033784.2 | 2 | 148 | ||
GenBank mRNA | AJ011413.1 | - | 2 | 135 | ||
AJ457860.1 | - | 2 | 102 | |||
AK010025.1 | - | 2 | 137 | |||
AK050248.1 | - | 2 | 137 | |||
AK131891.1 | - | 2 | 140 | |||
AK160487.1 | - | 2 | 140 | |||
BC024643.1 | - | 2 | 122 | 4242076 | ||
BC049971.1 | - | 2 | 122 | 4161986 | ||
BY131635.1 | - | 2 | 168 | |||
JX564908.1 | - | 2 | 102 | |||
JX564909.1 | - | 2 | 102 | |||
JX564910.1 | - | 2 | 102 |