Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alcam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009655.2 | NP_033785.1 | 8 | 1514 | ||
GenBank mRNA | AB003026.1 | - | 8 | 1176 | ||
AK030851.1 | - | 8 | 1515 | |||
AK031391.1 | - | 2590 | ||||
BC027280.1 | - | 8 | 1171 | 3501215 | ||
L25274.1 | - | 3 | 306 | |||
U95030.1 | - | 8 | 982 |