Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013467.3 | NP_038495.2 | 10 | 1082 | ||
GenBank mRNA | BC035322.1 | - | 10 | 1076 | ||
BC044729.2 | - | 10 | 1083 | |||
BC054386.1 | - | 10 | 1082 | 6477503 | ||
M74570.1 | - | 10 | 1096 | |||
S75713.1 | - | 10 | 1052 |