Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldoa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177307.1 | NP_001170778.1 | 9 | 1158 | ||
NM_001177308.1 | NP_001170779.1 | 8 | 1031 | |||
NM_007438.4 | NP_031464.1 | 8 | 1081 | |||
GenBank mRNA | AK002561.1 | - | 8 | 970 | ||
AK011238.1 | - | 4 | 491 | |||
AK145639.1 | - | 8 | 968 | |||
AK154295.1 | - | 8 | 964 | |||
AK162290.1 | - | 8 | 967 | |||
BC043026.1 | - | 8 | 957 | 5691975 | ||
BC050896.1 | - | 8 | 999 | 5715436 | ||
BC066218.1 | - | 8 | 977 | 30054842 | ||
BC066801.1 | - | 8 | 977 | 30072521 | ||
BC089495.1 | - | 8 | 1010 | 30312668 | ||
EF662059.1 | - | 8 | 1012 | |||
EF662060.1 | - | 8 | 962 | |||
Y00516.1 | - | 8 | 927 |
Gene Symbol: Aldoart2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277340.1 | NP_001264269.1 | 1 | 937 | ||
GenBank mRNA | AK006425.1 | - | 1 | 924 | ||
AK016920.1 | - | 1 | 904 | |||
EF662062.1 | - | 1 | 851 |