Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr1b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009658.3 | NP_033788.3 | 4, 5 | 517 | ||
GenBank mRNA | AK028079.1 | - | 4, 5 | 472 | ||
AK139250.1 | - | 4, 5 | 472 | |||
AK145392.1 | - | 4, 5 | 473 | |||
AK149861.1 | - | 4, 5 | 507 | |||
AK150197.1 | - | 4, 5 | 472 | |||
AK153949.1 | - | 4, 5 | 501 | |||
AK164682.1 | - | 4, 5 | 472 | |||
AK170663.1 | - | 4, 5 | 519 | |||
BC004725.1 | - | 4, 5 | 464 | 3582788 | ||
BC021655.1 | - | 4, 5 | 510 | 3708927 | ||
BC085310.1 | - | 4, 5 | 449 | 6485416 | ||
BY749711.1 | - | 4, 5 | 517 | |||
D32250.1 | - | 4, 5 | 458 | |||
L39795.1 | - | 4, 5 | 449 | |||
U29152.1 | - | 4, 5 | 468 |