Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alox12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007440.4 | NP_031466.2 | 4 | 562 | ||
GenBank mRNA | AK036898.1 | - | 4 | 560 | ||
AK036960.1 | - | 4 | 518 | |||
AK079232.1 | - | 4 | 563 | |||
AK087724.1 | - | 4 | 563 | |||
AK172216.1 | - | 4 | 560 | |||
AK172488.1 | - | 4 | 563 | |||
BC152329.1 | - | 4 | 567 | 40141477 | ||
BC152403.1 | - | 4 | 567 | 40141482 | ||
S77511.1 | - | 2 | 119 | |||
S80446.1 | - | 4 | 574 | |||
U04334.1 | - | 4 | 493 |