Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDIA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005313.4 | NP_005304.3 | 2 | 382 | ||
GenBank mRNA | AK075455.1 | - | 3 | 496 | ||
AK075481.1 | - | 3 | 283 | |||
BC014433.1 | - | 2 | 323 | 3542405 | ||
BC036000.1 | - | 2 | 336 | 4712175 | ||
BC071878.1 | - | 2 | 325 | 6380973 | ||
BG826781.1 | - | 2 | 383 | |||
D16234.1 | - | 2 | 301 | |||
D83485.1 | - | 2 | 256 | |||
EU668338.1 | - | 2 | 336 | |||
FJ224330.1 | - | 2 | 325 | |||
U42068.1 | - | 2 | 312 | |||
Z49835.1 | - | 2 | 340 |
Gene Symbol: PDIA3P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002305.1 | 1 | 497 | |||
GenBank mRNA | AK295985.1 | - | 1 | 382 | ||
BM555251.1 | - | 1 | 441 |