Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alox15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009660.3 | NP_033790.3 | 4, 5 | 582 | ||
GenBank mRNA | AK142371.1 | - | 4, 5 | 583 | ||
BC056625.1 | - | 4, 5 | 569 | 5005087 | ||
BC081546.1 | - | 4, 5 | 568 | 3671948 | ||
CT010263.1 | - | 4, 5 | 556 | |||
L34570.1 | - | 4, 5 | 566 | |||
U04331.1 | - | 4, 5 | 556 |