Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alox8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009661.4 | NP_033791.1 | 2 | 277 | ||
GenBank mRNA | AK028724.1 | - | 2 | 273 | ||
BC015253.1 | - | 2 | 277 | 4221815 | ||
U93277.1 | - | 2 | 243 | |||
Y14696.1 | - | 2 | 256 |