Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alox8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009661.4 | NP_033791.1 | 5 | 689 | ||
GenBank mRNA | AK028724.1 | - | 5 | 685 | ||
AK158553.1 | - | 3 | 563 | |||
BC015253.1 | - | 5 | 689 | 4221815 | ||
U93277.1 | - | 5 | 655 | |||
Y14696.1 | - | 5 | 668 |