Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDIA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005313.4 | NP_005304.3 | 3 | 461 | ||
GenBank mRNA | AK075455.1 | - | 4 | 575 | ||
AK075481.1 | - | 4 | 362 | |||
AK293249.1 | - | 2 | 134 | |||
BC014433.1 | - | 3 | 402 | 3542405 | ||
BC036000.1 | - | 3 | 415 | 4712175 | ||
BC071878.1 | - | 3 | 404 | 6380973 | ||
D16234.1 | - | 3 | 380 | |||
D83485.1 | - | 3 | 335 | |||
EU668338.1 | - | 3 | 415 | |||
FJ224330.1 | - | 3 | 404 | |||
U42068.1 | - | 3 | 391 | |||
Z49835.1 | - | 3 | 419 |