Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfer | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023040.3 | NP_075527.2 | 1, 2 | 331 | ||
GenBank mRNA | AB041561.1 | - | 1, 2 | 248 | ||
AF148688.1 | - | 1, 2 | 111 | |||
AK146579.1 | - | 1, 2 | 332 | |||
BC023941.1 | - | 1, 2 | 331 | 5342526 |