Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110505.1 | NP_001103975.1 | 7, 8 | 1096 | ||
NM_007446.2 | NP_031472.2 | 7, 8 | 1205 | |||
GenBank mRNA | AK029642.1 | - | 7, 8 | 1128 | ||
BC004698.1 | - | 3, 4 | 439 | |||
BC009121.1 | - | 7, 8 | 1079 | 3710941 | ||
J00356.1 | - | 6, 7 | 1049 | |||
V00717.1 | - | 7, 8 | 1094 | |||
V00719.1 | - | 7, 8 | 1205 | |||
V00720.1 | - | 7, 8 | 1096 |