Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRSF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098477.1 | NP_001091947.1 | 6 | 884 | ||
NM_002092.3 | NP_002083.3 | 6 | 1048 | |||
XM_005265681.3 | XP_005265738.1 | 6 | 1287 | |||
XM_011531897.2 | XP_011530199.1 | 6 | 713 | |||
GenBank mRNA | AK055499.1 | - | 7 | 824 | ||
AK056891.1 | - | 7 | 794 | |||
AK090755.1 | - | 7 | 828 | |||
AK094806.1 | - | 6 | 853 | |||
AK095799.1 | - | 7 | 814 | |||
AK097055.1 | - | 7 | 894 | |||
AK124124.1 | - | 7 | 965 | |||
AK127161.1 | - | 7 | 923 | |||
AK294612.1 | - | 6 | 748 | |||
AK296287.1 | - | 7 | 883 | |||
AK298883.1 | - | 6 | 706 | |||
U07231.1 | - | 7 | 858 |