Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ank1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110783.3 | NP_001104253.1 | 3 | 590 | ||
NM_001277280.2 | NP_001264209.1 | 3 | 590 | |||
NM_001277281.2 | NP_001264210.1 | 3 | 311 | |||
NM_001277284.2 | NP_001264213.1 | 3 | 311 | |||
NM_001277286.2 | NP_001264215.1 | 3 | 576 | |||
NM_001277289.2 | NP_001264218.1 | 3 | 576 | |||
NM_031158.4 | NP_112435.2 | 3 | 576 | |||
GenBank mRNA | AK147278.1 | - | 3 | 545 | ||
AK147483.1 | - | 3 | 576 | |||
AK147597.1 | - | 3 | 524 | |||
AK158131.1 | - | 3 | 583 | |||
BC079910.1 | - | 3 | 282 | 30664024 | ||
BC138029.1 | - | 3 | 310 | 8861048 | ||
BC145102.1 | - | 3 | 310 | |||
BC171944.1 | - | 3 | 310 | 9054230 | ||
CJ101195.1 | - | 3 | 312 | |||
M84756.1 | - | 3 | 288 | |||
X69063.1 | - | 3 | 560 |