Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankfy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009671.5 | NP_033801.4 | 9, 10 | 1193 | ||
GenBank mRNA | AB098329.1 | - | 9, 10 | 1191 | ||
BC139231.1 | - | 9, 10 | 1204 | 8862253 | ||
BC145583.1 | - | 9, 10 | 1204 | 9054149 | ||
CV557421.1 | - | 2, 3 | 220 |