Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013470.2 | NP_038498.2 | 6 | 559 | ||
GenBank mRNA | AJ001633.1 | - | 6 | 542 | ||
AK013850.1 | - | 4 | 312 | |||
AK090055.1 | - | 6 | 577 | |||
AK150922.1 | - | 6 | 560 | |||
AK150951.1 | - | 6 | 527 | |||
AK151771.1 | - | 6 | 519 | |||
AK152837.1 | - | 6 | 486 | |||
AK152863.1 | - | 6 | 547 | |||
AK166389.1 | - | 6 | 531 | |||
AK168396.1 | - | 6 | 455 | |||
AK168507.1 | - | 6 | 487 | |||
AK169113.1 | - | 6 | 435 | |||
AK169423.1 | - | 6 | 441 | |||
BC090634.1 | - | 6 | 553 | 30942562 | ||
BC141167.1 | - | 6 | 513 | 9055779 | ||
CT010280.1 | - | 4 | 342 |