Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013471.2 | NP_038499.2 | 12 | 1002 | ||
GenBank mRNA | AK132293.1 | - | 13 | 1044 | ||
AK150486.1 | - | 12 | 963 | |||
AK150614.1 | - | 12 | 949 | |||
AK151054.1 | - | 12 | 949 | |||
AK151236.1 | - | 12 | 981 | |||
AK167338.1 | - | 12 | 1003 | |||
AK168390.1 | - | 12 | 974 | |||
AK168487.1 | - | 12 | 983 | |||
AK168917.1 | - | 11 | 1007 | |||
AK170447.1 | - | 12 | 948 | |||
BC055871.1 | - | 12 | 962 | 4947415 | ||
U72941.1 | - | 12 | 973 | |||
U95371.1 | - | 13 | 1661 |