Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110794.1 | NP_001104264.1 | 8 | 953 | ||
NM_009674.3 | NP_033804.2 | 8 | 1068 | |||
GenBank mRNA | AK032013.1 | - | 8 | 844 | ||
AK077579.1 | - | 8 | 761 | |||
AK134589.1 | - | 9 | 838 | |||
AK149654.1 | - | 8 | 760 | |||
AK151438.1 | - | 8 | 762 | |||
AK161613.1 | - | 8 | 762 | |||
AK167588.1 | - | 8 | 736 | |||
AK169199.1 | - | 8 | 735 | |||
AK178561.1 | - | 5 | 386 | |||
AK203311.1 | - | 3 | 206 | |||
BC008997.1 | - | 8 | 752 | 3157926 | ||
L13129.1 | - | 8 | 755 |