Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aoc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009675.2 | NP_033805.1 | 1 | 1385 | ||
GenBank mRNA | AF054831.1 | - | 1 | 1400 | ||
AF115411.1 | - | 1 | 1216 | |||
BC080857.1 | - | 1 | 1224 | 6854465 |