Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap1m2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110300.1 | NP_001103770.1 | 10 | 1280 | ||
NM_009678.2 | NP_033808.2 | 10 | 1274 | |||
GenBank mRNA | AF067146.1 | - | 10 | 1248 | ||
AK010478.1 | - | 10 | 1259 | |||
AK041933.1 | - | 2 | 52 | |||
AK082674.1 | - | 1137 | ||||
AK160161.1 | - | 10 | 1245 | |||
BC003704.1 | - | 10 | 1213 | 3488231 |