Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077264.1 | NP_001070732.1 | 5 | 631 | ||
NM_007458.2 | NP_031484.1 | 5 | 631 | |||
GenBank mRNA | AK135424.1 | - | 5 | 560 | ||
AK155515.1 | - | 5 | 567 | |||
BC031433.1 | - | 5 | 581 | 4481643 | ||
CA327188.1 | - | 5 | 637 |