Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apbb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201413.1 | NP_001188342.1 | 15 | 2453 | ||
NM_001201414.1 | NP_001188343.1 | 14 | 2456 | |||
NM_001201415.1 | NP_001188344.1 | 14 | 2390 | |||
NM_001201416.1 | NP_001188345.1 | 5 | 556 | |||
NM_009686.2 | NP_033816.1 | 16 | 2459 | |||
GenBank mRNA | AF133665.1 | - | 1 | 31 | ||
AK004792.1 | - | 16 | 2313 | |||
AK153260.1 | - | 14 | 2422 | |||
AK167649.1 | - | 15 | 2318 | |||
AK169959.1 | - | 14 | 2328 | |||
AK172405.1 | - | 5 | 560 | |||
BC042797.1 | - | 6 | 598 | |||
BC076587.1 | - | 14 | 2236 | 30550087 |