Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apbb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201413.1 | NP_001188342.1 | 16, 17 | 2684 | ||
NM_001201414.1 | NP_001188343.1 | 15, 16 | 2687 | |||
NM_001201415.1 | NP_001188344.1 | 15, 16 | 2621 | |||
NM_001201416.1 | NP_001188345.1 | 6, 7 | 787 | |||
NM_009686.2 | NP_033816.1 | 17, 18 | 2690 | |||
GenBank mRNA | AF133665.1 | - | 2, 3 | 262 | ||
AK004792.1 | - | 17, 18 | 2544 | |||
AK153260.1 | - | 15, 16 | 2653 | |||
AK167649.1 | - | 16, 17 | 2549 | |||
AK169959.1 | - | 15, 16 | 2559 | |||
AK172405.1 | - | 6, 7 | 791 | |||
BC042797.1 | - | 7, 8 | 829 | |||
BC076587.1 | - | 15, 16 | 2467 | 30550087 |