Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Speg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085370.1 | NP_001078839.1 | 7 | 2091 | ||
NM_001085371.1 | NP_001078840.1 | 10 | 2558 | |||
NM_001173477.1 | NP_001166948.1 | 4 | 571 | |||
NM_007463.4 | NP_031489.4 | 9 | 2991 | |||
GenBank mRNA | AF215896.1 | - | 9 | 2991 | ||
AK147475.1 | - | 7 | 2096 | |||
AK164360.1 | - | 10 | 2537 | |||
BC048698.1 | - | 4 | 468 | |||
BC062643.1 | - | 5 | 683 | |||
CK622939.1 | - | 4 | 518 | |||
U57098.1 | - | 3 | 398 |