Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xiap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301639.1 | NP_001288568.1 | 3, 4 | 1485 | ||
NM_001301641.1 | NP_001288570.1 | 3, 4 | 1272 | |||
NM_009688.3 | NP_033818.2 | 4, 5 | 1357 | |||
GenBank mRNA | AK147447.1 | - | 3, 4 | 1487 | ||
BB650856.1 | - | 3, 4 | 311 | |||
BB663325.1 | - | 2, 3 | 222 | |||
BC138528.1 | - | 2, 3 | 1280 | 8861548 | ||
BC145861.1 | - | 2, 3 | 1280 | 40131164 | ||
U36842.1 | - | 3, 4 | 1189 | |||
U88990.1 | - | 2, 3 | 1649 |