Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aplp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102455.1 | NP_001095925.1 | 17 | 2295 | ||
NM_001102456.1 | NP_001095926.1 | 17 | 2163 | |||
NM_009691.2 | NP_033821.1 | 16 | 2127 | |||
GenBank mRNA | AK088391.1 | - | 4 | 518 | ||
AK149977.1 | - | 17 | 2296 | |||
AK150020.1 | - | 17 | 2296 | |||
AK150478.1 | - | 9 | 1027 | |||
AK151708.1 | - | 9 | 1027 | |||
AK152787.1 | - | 9 | 1028 | |||
AK153087.1 | - | 9 | 1027 | |||
BC026491.1 | - | 7 | 579 | |||
BC051999.1 | - | 17 | 2141 | 5716148 | ||
BC052396.1 | - | 16 | 2099 | 5719918 | ||
M97216.1 | - | 12 | 1487 | |||
U15571.1 | - | 17 | 2271 | |||
Z22592.1 | - | 16 | 2127 | |||
Z27070.1 | - | 16 | 2125 |