Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apobec2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009694.3 | NP_033824.1 | 2 | 520 | ||
GenBank mRNA | AF161699.1 | - | 2 | 521 | ||
AK009047.1 | - | 2 | 418 | |||
BC027530.1 | - | 2 | 430 | 1433092 |