Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apoc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110009.2 | NP_001103479.1 | 3 | 190 | ||
NM_007469.5 | NP_031495.1 | 2 | 312 | |||
GenBank mRNA | AK011358.1 | - | 2 | 227 | ||
BC019398.1 | - | 3 | 165 | 5124986 | ||
BC035445.1 | - | 3 | 177 | |||
BC094638.1 | - | 2 | 159 | 875610 | ||
BY087738.1 | - | 3 | 191 | |||
CB947285.1 | - | 2 | 279 |