Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000178.3 | NP_000169.1 | 6 | 637 | ||
NM_001322494.1 | NP_001309423.1 | 6 | 704 | |||
NM_001322495.1 | NP_001309424.1 | 6 | 1466 | |||
GenBank mRNA | AK293144.1 | - | 6 | 528 | ||
AK298292.1 | - | 4 | 815 | |||
AK311567.1 | - | 4 | 722 | |||
AK312492.1 | - | 6 | 617 | |||
BC007927.1 | - | 6 | 624 | 3946317 | ||
EU668322.1 | - | 6 | 624 | |||
EU794618.1 | - | 6 | 624 | |||
KU178105.1 | - | 5 | 538 | |||
U34683.1 | - | 6 | 578 |