Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aprt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009698.2 | NP_033828.2 | 5 | 793 | ||
GenBank mRNA | AK002350.1 | - | 5 | 746 | ||
AK153201.1 | - | 5 | 758 | |||
BC005667.1 | - | 5 | 731 | 3152733 | ||
M11310.1 | - | 2989 |