Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aqp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009699.3 | NP_033829.3 | 4 | 1280 | ||
GenBank mRNA | AK002689.1 | - | 4 | 1279 | ||
AK142567.1 | - | 4 | 1281 | |||
BC019966.1 | - | 4 | 1264 | 4222942 |